Order Entry
Austria
ContactUsLinkComponent
Copper(II) hexafluoroacetylacetonate hydrate
Irritant
Copper(II) hexafluoroacetylacetonate hydrate
Catalog # 14505.03
CAS Number:  
Irritant
Copper(II) hexafluoroacetylacetonate hydrate
Catalog # 14505.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
 :477.64 g/mol (anhy)
Boiling Pt:220 °C (dec.)
Melting Pt:∼ 100 °C
EINECS:238-850-1
 :GHS07
Storage Temperature:Ambient
CAS Number:155640-85-0
 :14781-45-4
Formula:C₁₀H₂CuF₁₂O₄·xH₂O
MDL Number:MFCD00151019