Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')chromium(III), Sigma-Aldrich®
Catalog # SIAL468223-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:601.8 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:14434-47-0
Formula:C₃₃H₅₇CrO₆
MDL Number:MFCD00058919