Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)calcium ≥99.99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:118448-18-3
:
Formula:C₂₂H₃₈CaO₄
MDL Number:MFCD00064757
Boiling Pt:205 °C (0.1 mmHg) (subl.)
Melting Pt:202…206 °C
:406.57 g/mol
Storage Temperature:Ambient
Related Information
Frequently Bought Together