Order Entry
Austria
ContactUsLinkComponent
Gel filtration media, Sephadex™ G-100
Gel filtration media, Sephadex™ G-100
Supplier:  Cytiva
Certificates
undefined
Gel filtration media, Sephadex™ G-100
Supplier:  Cytiva
Certificates

Specifications

  • pH range:
    2 - 10
  • Matrix:
    Cross-linked dextran
  • Max. pressure:
    0,096 bar

Specifications

Related Information

Frequently Bought Together