Order Entry
Austria
ContactUsLinkComponent
Gel filtration media, Sephadex™ G-10
Gel filtration media, Sephadex™ G-10
Supplier:  Cytiva
Certificates
undefined
Gel filtration media, Sephadex™ G-10
Supplier:  Cytiva

Specifications

  • pH range:
    2 - 13
  • Matrix:
    Cross-linked dextran
  • Fractionation range:
    <7 x 10² dextrans, <7 x 10² globular proteins
  • Storage temperature:
    4 - 30 °C

Specifications

Related Information

Frequently Bought Together