TLC plates, cellulose MN 400, AVICEL®
Lieferant: MACHEREY-NAGEL
Zertifikate
Über diesen Artikel
These TLC plates and sheets are prepared by hydrolysis of high purity cellulose with HCl. They are used for carboxylic acids, lower alcohols, urea and purine derivatives.
- Alternative selectivity to silica 60
Celluolse MN 400 (AVICEL®): Microcrystalline; average degree of polymerisation 40 to 200.
Frequently Bought Together