TLC plates, aluminium oxide, Alox
TLC plates and sheets are used for the separations of alkaloids, steroids, aliphatic and aromatic compounds.
- Alternative selectivity to silica 60
Aluminium oxide: Specific surface (BET) ~ 200 m²/g
Indicator: Manganese activated zinc silicate
Frequently Bought Together