ContactUsLinkComponent
Reusable earplugs, 1261/1271, Comets®
Reusable earplugs, 1261/1271, Comets®
Catalog # 111-1163
Supplier:  Moldex
undefined
Reusable earplugs, 1261/1271, Comets®
Catalog # 111-1163
Supplier:  Moldex
Specifications
  • Colour:
    Light green
  • Description:
    Earplugs, with cord, in pocket case
  • Size:
    U*
  • Environmentally Preferable:
  • Pk:
    50 PAIR
Specifications
About this item
These reusable earplugs provide protection against lower noise levels. They are designed to keep the user in touch with the environment.
  • Air bubble in the tip and soft material provide a comfortable seal
  • Plugs made of soft durable material – Washable and hygienic
  • Come with handy PocketPak storage container
SNR: 25 dB
Certifications: EN 352-2:2002
Caution: * One size fits all.