HPLC columns, Accucore™ Phenyl-X
Supplier: Thermo Fisher Scientific
Certificates
About this item
Achieve fast, high-resolution separations of aromatic compounds using Thermo Scientific™ Accucore Phenyl-X columns.
Rugged 2,6 µm solid core particles enable fast, high efficiency separations at low backpressures. The Phenyl-X alkyl aromatic bonded phase offers unique reversed-phase shape selectivity with high aromatic selectivity and is compatible with highly aqueous mobile phases. Robust bonding technology and automated packing procedures ensure excellent reproducibility and long column lifetimes.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm int.Ø require the 1 mm UNIGUARD holder (10046-746), 2,1/3 mm int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4,6 mm int.Ø require the 4/4,6 mm UNIGUARD holder (10046-744).
Frequently Bought Together