Graduated pipettes, BLAUBRAND®, USP, type 2, Class AS, total delivery
: Brand
AR-Glas®
- Calibrated to deliver (TD, Ex), nominal volume at the top
- Marks and inscriptions in high contrast blue enamel
- Waiting time 5 seconds
- Suitable for cotton plugs
- Incl. USP batch certificate, on request also available with USP individual certificate
DIN EN ISO 835
Length: 360 ±10 mm
: DIN EN ISO 835. DE-M marking.
: * Error limit USP: ±0,01 ml.
Frequently Bought Together