Order Entry
Austria
ContactUsLinkComponent
Fibrillar collagen cell culture inserts, Corning® BioCoat®
Fibrillar collagen cell culture inserts, Corning® BioCoat®
Catalog # 734-0154
Supplier:  Corning
   Low Manufacturing Impact
Fibrillar collagen cell culture inserts, Corning® BioCoat®
Catalog # 734-0154
Supplier:  Corning
   Low Manufacturing Impact

Specifications

  • Pore size:
    0,1 µm
  • Description:
    Inserts, packed in two 24-well plates
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 - Bedford
  • Pk:
    24

Specifications

About this item

Corning BioCoat cell culture inserts are pre-coated with extracellular matrix proteins for applications requiring a protein-coated cell surface, such as cell adhesion, growth, invasion, migration and/or differentiation.

  • Storage: 2 to 8 °C
  • Shelf life: >= 3 months from date of shipment