Order Entry
Austria
ContactUsLinkComponent
Dispensing pumps, '30'
Dispensing pumps, '30'
Catalog # MURRP514/14
Supplier:  MURRAY S
Dispensing pumps, '30'
Catalog # MURRP514/14
Supplier:  MURRAY S

Specifications

  • Flow rate:
    0,5 ml/stroke
  • Description:
    Dispensing pump ‘30’, white, neck 38 mm
  • Environmentally Preferable:
  • Pk:
    20

Specifications

About this item

Dispensing pump is suitable for lotions, creams and gels.

  • Locking device for shipment