Containers, wide mouth, with snap-on cap
Supplier: MURRAY S
Certificates
About this item
PP, natural.
- Caps (PE) have tear-off strip for immediate visual confirmation of product integrity
- Suitable for silk screen printing, and dry offset printing in up to 4 colours plus a clear lacque
Wide neck tamper evident containers suitable for tablets and powder.
Delivery information: Supplied in standard carton quantities.
Frequently Bought Together