Single channel micro pipettes, mechanical, fixed volume, Acura® manual 815
Lieferant: Socorex Isba
Zertifikate
Über diesen Artikel
These pipettes show a very stable performance and provide consistent results for any analytical or routine tests.
- Ergonomic shape, lightweight and soft plunger action
- Volume setting is performed smoothly and precisely by turning push button; click-stops and free rotating caps prevent any unwanted volume alteration
- Display is located at the front and therefore always visible at all times during pipetting
- Justip™ tip ejector button
- Autoclavable
Frequently Bought Together