Wound cleansers
Supplier: ORKLA CARE
Certificates
About this item
Wound cleansers do away with cotton wool and bottles of antiseptic. These sterile, non woven tissues, which are saturated in a wound cleansing fluid (0,9% NaCl), are intended for the mechanical cleansing of minor wounds when clean tap water is not available.
- Tissues are packed in sealed sachets, which prevent leakage and guarantee sterility
- Savett sachets are packed in a refill box
- Salvequick cleansers fit into first-aid-stations
Frequently Bought Together