ContactUsLinkComponent
Deionizers with nylon resin canisters, behropur®
Deionizers with nylon resin canisters, behropur®
Supplier:  BEHR
Certificates
undefined
Deionizers with nylon resin canisters, behropur®
Supplier:  BEHR
Specifications
  • Catalog No:
  • BEHRB00223471
  • BEHRB00223469
  • BEHRB00223473
  • Model:
  • B22dN
  • B10dN
  • B45dN
  • Capacity:
  • 2400 L
  • 1200 L
  • 5500 L
  • Max. flow rate:
  • 500 l/h
  • 300 l/h
  • 800 l/h
  • Ø×H:
  • 210×1120 mm
  • 210×680 mm
  • 260×1250 mm