Orders LinkContactUsLinkComponent
Bath fluids
Bath fluids
Supplier:   HUBER
Bath fluids
Supplier:  HUBER
Certificates
Bath fluids
23 Options Available Below
Product Details & Documents
Thermal fluids with a wide range of operating temperatures and good thrmodynamic properties for use in circulators and heating systems where water is not appropriate.
All viscosities values are dynamic visosities measured at 25 °C except DW-Therm® which was measured at 20 °C.
Caution: * –20...+235 °C for closed system applications
Use biocides safely. Always read the label and product information before use
Product Family Options
Thermal fluids for closed systems, eg Unistats
Product Information
  • Type
    Temperature range
    Flash point
    Viscosity
    Pk
    Availability
    Price
  • DW-Therm HT®
    20...340 °C
    ~190 °C
    32 mm²/s
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 725,00 €
    725,00 €
    1 * 10 L
       725,00 €
  • DW Therm®
    −90...+200 °C
    101 °C
    2,0 -2,2 mm²/s
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 1 040,00 €
    1 040,00 €
    1 * 10 L
       1 040,00 €
  • DW-Therm HT®
    20...340 °C
    ~190 °C
    32 mm²/s
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 420,00 €
    420,00 €
    1 * 5 L
       420,00 €
Thermal fluids for open systems
Product Information
  • Type
    Temperature range
    Flash point
    Viscosity
    Pk
    Availability
    Price
  • SilOil
    20...275 °C
    300 °C
    50 mm²/s
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 805,00 €
    805,00 €
    1 * 10 L
       805,00 €
  • SilOil
    −40...+165 °C
    >170 °C
    10 mm²/s
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 805,00 €
    805,00 €
    1 * 10 L
       805,00 €
  • SilOil
    −20...+195* °C
    >240 °C
    20 mm²/s
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 805,00 €
    805,00 €
    1 * 10 L
       805,00 €
  • SilOil
    −90...+55 °C
    >90 °C
    3 mm²/s
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 1 810,00 €
    1 810,00 €
    1 * 10 L
       1 810,00 €
  • SilOil
    −90...+55 °C
    >90 °C
    3 mm²/s
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 915,00 €
    915,00 €
    1 * 5 L
       915,00 €
  • SilOil
    −20...+195* °C
    >240 °C
    20 mm²/s
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 450,00 €
    450,00 €
    1 * 5 L
       450,00 €
  • MinOil
    20...190 °C
    >184 °C
    40 mm²/s
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 230,00 €
    230,00 €
    1 * 5 L
       230,00 €
  • MinOil
    20...190 °C
    >184 °C
    40 mm²/s
    20 L
    Log in for availability
    Log in for availability
    1 * 20 L
    1 * 20 L - 690,00 €
    690,00 €
    1 * 20 L
       690,00 €
  • SilOil
    20...275 °C
    300 °C
    50 mm²/s
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 450,00 €
    450,00 €
    1 * 5 L
       450,00 €
  • SilOil
    −40...+165 °C
    >170 °C
    10 mm²/s
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 450,00 €
    450,00 €
    1 * 5 L
       450,00 €
  • SilOil
    −60...+115 °C
    >120 °C
    5 mm²/s
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 450,00 €
    450,00 €
    1 * 5 L
       450,00 €
  • SilOil
    −60...+115 °C
    >120 °C
    5 mm²/s
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 805,00 €
    805,00 €
    1 * 10 L
       805,00 €
  • SilOil
    −80...+55 °C
    >62 °C
    3 mm²/s
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 485,00 €
    485,00 €
    1 * 5 L
       485,00 €
  • SilOil
    −80...+55 °C
    >62 °C
    3 mm²/s
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 840,00 €
    840,00 €
    1 * 10 L
       840,00 €
  • SilOil
    −80...+100 °C
    >121 °C
    3 mm²/s
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 950,00 €
    950,00 €
    1 * 5 L
       950,00 €
  • SilOil
    −80...+100 °C
    >121 °C
    3 mm²/s
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 1 820,00 €
    1 820,00 €
    1 * 10 L
       1 820,00 €
  • SynOil
    −10...+120 °C
    125 °C
    8 mm²/s
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 505,00 €
    505,00 €
    1 * 10 L
       505,00 €
Antifreeze - cooling fluids
Product Information
  • Type
    Temperature range
    Flash point
    Viscosity
    Pk
    Availability
    Price
  • Ethylene glycol
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 330,00 €
    330,00 €
    1 * 10 L
       330,00 €
  • Ethylene glycol
    50 L
    Log in for availability
    Log in for availability
    1 * 50 L
    1 * 50 L - 1 200,00 €
    1 200,00 €
    1 * 50 L
       1 200,00 €
  • Ethylene glycol
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 200,00 €
    200,00 €
    1 * 1 ST
       200,00 €
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.