Digestion system, behrotest®
Supplier: BEHR
Jump to:
About this item
Digestion apparatus for crude fibre or hydrolysis.
: Includes serial heating unit with individually adjustable heating places, cooling water distribution with condenser rack and hoses, beakers (600 ml), cold fingers.
3 Options Available Below
Product Details & Documents
Digestion apparatus for crude fibre or hydrolysis.
: Includes serial heating unit with individually adjustable heating places, cooling water distribution with condenser rack and hoses, beakers (600 ml), cold fingers.
Product Family Options
Product Information
- TypeAvailabilityPrice
- BEHRB00218430Hydrolysis unit, complete for one sample
- BEHRB00218446Complete crude fibre digestion unit with 4 sample positionsComplete crude fibre digestion unit with 4 sample positionsEXR 4Log in for availability1 * 1 ST1 956,36 €
- BEHRB00218448Complete crude fibre digestion unit with 6 sample positionsComplete crude fibre digestion unit with 6 sample positionsEXR 6Log in for availability1 * 1 ST2 435,00 €