Watch glasses
Supplier: MURRAY S
Certificates
About this item
Made of glass.
- Good chemical and physical properties
- Use in applications that do not involve high thermal stresses
- Quick and easy to clean
With ground edges for covering beakers.
Frequently Bought Together