Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Staining tile

12, Ø×H: 15×1,5 mm, 89×57×4,5 mm
  MARI1800002
 :   MARIENFELD
Staining tile
  MARI1800002
Staining tile
Staining tile

12, Ø×H: 15×1,5 mm, 89×57×4,5 mm

 
226,42 €
   
1 * 12 ST
 
 
  • No. of wells:
    12
  • W×D×H:
    89×57×4,5 mm
  • Well size:
    Ø×H: 15×1,5 mm