Orders LinkContactUsLinkComponent
Multiple or single cuvette segments
Multiple or single cuvette segments
Anbieter:   KARTELL
Multiple or single cuvette segments
Anbieter:  KARTELL
Certificates
Multiple or single cuvette segments
Multiple or single cuvette segments
2 Options Available Below
Product Details & Documents
These twelve position cuvettes segments have been specially designed for use with the Cobas Mira® analyser.
Packaging: In cases of bulk segments (450) or 30 pre-racked segments (15).
Product Family Options
Product Information
  • Description
    Material
    Verfügbarkeit
    Preis
  • Cuvettes in pre-racked segments (15)
    PMMA
    Log in for availability
    Log in for availability
    1 * 450 ST
    1 * 450 ST - 734,00 €
    734,00 €
    1 * 450 ST
       734,00 €
  • Cuvettes in bulk segments
    PMMA
    Log in for availability
    Log in for availability
    1 * 450 ST
    1 * 450 ST - 679,00 €
    679,00 €
    1 * 450 ST
       679,00 €