Order Entry
Austria
ContactUsLinkComponent
Silver billet electrode, Orion™
Silver billet electrode, Orion™
Catalog # ORIO9780SC
Supplier:  Thermo Orion
undefined
Silver billet electrode, Orion™
Catalog # ORIO9780SC
Supplier:  Thermo Orion

Specifications

  • Connection:
    Screw cap
  • Ø×L:
    12×150 mm
  • For:
    Halide titrations
  • Description:
    Silver billet combination electrode
  • Environmentally Preferable:
  • Diaphr.:
    Ceramic
  • Electrolyte:
    4 M KCl w/Ag/AgCl
  • Pk:
    1
  • Temp.:
    0...+80 °C

Specifications

About this item

These electrodes are a combination of a silver billet-indicating electrode along with a silver billet half-cell. Ideal for obtaining an accurate determination of potentiometric halides with dual electrodes.

Internal reference: Ag/AgCl