Orders LinkContactUsLinkComponent
Conductivity standards
Conductivity standards
Supplier:   Cole-Parmer
Conductivity standards
Supplier:  Cole-Parmer
Certificates
Conductivity standards
2 Options Available Below
Product Details & Documents
Standard solutions are used for calibrating conductivity cells.
Product Family Options
Conductivity standards
Product Information
  • Conductivity
    Pk
    Availability
    Price
  • JENW16156-59
    Conductivity standard (500 ml)
    Conductivity standard (500 ml)
    1413 µS
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 83,40 €
    83,40 €
    1 * 1 ST
       83,40 €
TDS standards
Product Information
  • Conductivity
    Pk
    Availability
    Price
  • JENW16156-81
    1382 ppm TDS calibration standard
    1382 ppm TDS calibration standard
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 105,00 €
    105,00 €
    1 * 500 mL
       105,00 €