ContactUsLinkComponent
Injection and aspiration filter, Sterifix® Pury
Injection and aspiration filter, Sterifix® Pury
Catalog # 612-0192
Supplier:  B. Braun
undefined
Injection and aspiration filter, Sterifix® Pury
Catalog # 612-0192
Supplier:  B. Braun
Specifications
  • Description:
    Injection and aspiration filters, 5 μm, with Lock fitting
  • Pk:
    100
Specifications
About this item
Injection and aspiration filter for the prevention of particulate contamination during injection and aspiration of drugs.