Vials, pathological
Supplier: MURRAY S
Certificates
About this item
Soda glass.
Tubular vials complete with fitted white wadless polypropylene cap.
Frequently Bought Together
Packaging Bags with Closure
Supplier: BURKLE
Screw caps for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
Grade 2V qualitative filter papers, fluted, Whatman™
Supplier: Whatman products (Cytiva)
VWR®, Chemical Resistant Gloves, Nitrile, Flock-Lined
Supplier: VWR Collection
Titration beakers
Supplier: Mettler - Toledo
Transfer pipettes, Falcon®
Supplier: Corning
VWR®, Conical Sample Containers, with Snap-On Lids
Supplier: VWR Collection
Weighing dishes, aluminium
Supplier: HEATHROW
BD Emerald™ Syringes
Supplier: BD Medical
Bottle top vacuum filtration systems, Nalgene®
Supplier: Thermo Fisher Scientific
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Sodium hydroxide ≥98%, pellets, EMPROVE® ESSENTIAL Ph. Eur., BP, FCC, JP, NF, E524, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
VWR®, Disposable Weighing Boats
Supplier: VWR Collection
Sporicide, Contec® Sterile Prochlor
Supplier: CONTEC
Thymolphthalein, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. indicator
Supplier: VWR Chemicals
Reagents for water determination by the Coulometric Titration Karl Fischer method, Hydranal™
Supplier: Honeywell Chemicals
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
Bottle top vacuum filtration systems
Supplier: SciLabware