Gel permeation chromatography columns, PLgel MiniMIX
Supplier: VARIAN
Certificates
About this item
These narrow bore columns offer high performance, excellent solvent compatibility and mechanical stability that can be used with conventional GPC equipment.
- High performance that uses about 70% less solvent and save money
- Store less solvent and increase operator safety
In order to maintain the same linear velocity through the column, the volumetric flow rate should be reduced to 0,3 ml/min in line with the column cross sectional area that resulting in significantly lower solvent consumption. Sample loading should also be scaled down in line with reduced column volume and system dead volume should be minimised to avoid excessive band broadening.
Frequently Bought Together