Orders LinkContactUsLinkComponent
Funnels, mini
Funnels, mini
Supplier:   BOCHEM
Funnels, mini
Supplier:  BOCHEM
Certificates
Funnels, mini
4 Options Available Below
Product Details & Documents
Mini funnel, made of 18/10 stainless steel.
  • Excellent temperature resistance
  • Good chemical resistance
  • Easy to clean
Product Family Options
Product Information
  • Length
    Funnel O-Ø
    Availability
    Price
  • BOCH8858
    outlet Ø 10 mm
    outlet Ø 10 mm
    50 mm
    31 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,20 €
    4,20 €
    1 * 1 ST
       4,20 €
  • BOCH8857
    outlet Ø 8 mm
    outlet Ø 8 mm
    50 mm
    29 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,20 €
    4,20 €
    1 * 1 ST
       4,20 €
  • BOCH8856
    outlet Ø 6 mm
    outlet Ø 6 mm
    50 mm
    27 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,20 €
    4,20 €
    1 * 1 ST
       4,20 €
  • BOCH8855
    outlet Ø 4 mm
    outlet Ø 4 mm
    50 mm
    27 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,20 €
    4,20 €
    1 * 1 ST
       4,20 €