Orders LinkContactUsLinkComponent
Forceps
Forceps
Supplier:   MURRAY S
Forceps
Supplier:  MURRAY S
Certificates
Forceps
2 Options Available Below
Product Details & Documents
Stainless steel. Dissecting forceps with guide pin, which helps to keep the fine tips aligned.
  • Sharp, narrow flutes
  • Curved
Product Family Options
Product Information
  • Type
    Length
    Availability
    Price
  • Sharp, curved
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7,50 €
    7,50 €
    1 * 1 ST
       7,50 €
  • Sharp, curved
    115 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,50 €
    4,50 €
    1 * 1 ST
       4,50 €