HPLC columns, NUCLEOSIL® C8
Supplier: MACHEREY-NAGEL
Certificates
About this item
NUCLEOSIL® C8 and C8 ec are octyl modified silica phases for HPLC.
- Robust spherical silica gel
- Weakly hydrophobic, polar, endcapped or non-endcapped phases for RP chromatography
- Separation mechanism based on hydrophobic (van der Waals) and slight or noticeable residual silanol interactions
EC guard columns are used for analytical EC columns with the Column Protection System guard column holder (554-5851).
Specifications
- Catalog No:
- MANA720990.46
- MANA720001.40
- MANA720735.46
- MANA720071.46
- MANA720052.46
- MANA720001.46
- MANA720071.40
- MANA720013.40
- MANA720050.40
- 554-5805
- MANA720013.46
- MANA720050.46
- MANA721194.30
- MANA720062.46
- 554-5800
- MANA721095.30
- MANA720022.40
- MANA720022.46
- MANA720052.40
- MANA720214.46
- MANA721093.30
- MANA720017.46
- MANA721061.30
- Carbon load:
- 8,5% (C8, 100 Å)
- 8,5% (C8, 100 Å)
- 6,5% (120 Å)
- 6,5% (120 Å)
- 6,5% (120 Å)
- 8,5% (C8, 100 Å)
- 6,5% (120 Å)
- 8,5% (C8, 100 Å)
- 6,5% (120 Å)
- 9% (C8 ec, 100 Å)
- 8,5% (C8, 100 Å)
- 6,5% (120 Å)
- 8,5% (C8, 100 Å)
- 3% (300 Å)
- 9% (C8 ec, 100 Å)
- 6,5% (120 Å)
- 8,5% (C8, 100 Å)
- 8,5% (C8, 100 Å)
- 6,5% (120 Å)
- 6,5% (120 Å)
- 6,5% (120 Å)
- 8,5% (C8, 100 Å)
- 3% (300 Å)
- Endcapped:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Yes
- No
- No
- No
- No
- Yes
- No
- No
- No
- No
- No
- No
- No
- No
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- USP code:
- L7, L7
- L7, L7
- L7, L7
- L7, L7
- L7, L7
- L7, L7
- L7, L7
- L7, L7
- L7, L7
- L7, L7
- L7, L7
- L7, L7
- L7, L7
- L7, L7
- L7, L7
- L7, L7
- L7, L7
- L7, L7
- L7, L7
- L7, L7
- L7, L7
- L7, L7
- L7, L7
Specifications
Frequently Bought Together