ContactUsLinkComponent
Repetitive pipette, manual, AutoRep™
  RAINAR-M2
 :  RAININ
Repetitive pipette, manual, AutoRep™
  RAINAR-M2
 :  RAININ
  • Volume:
    10 - 5000 µl
  • Description:
    AutoRep™ M, manual dispenser
  • Model:
    AutoRep™ M
  • Tip:
    Encode syringe tips; 0,5 - 50 ml
  • Pk:
    1
This manual repeater pipette can be used with Rainin Encode™ or Brand® PD-Tip® syringes or compatible syringes from other manufacturers.
  • Precise and accurate
  • Saves time and effort, speeding up long pipetting serie
  • Lightweight, ergonomic design virtually eliminates all forces that contribute to repetitive strain injuries experienced in multi-dispense pipetting
Encode syringe tips