Ion exchange HPLC columns, Bio
: Agilent
These HPLC columns are packed with polymeric, nonporous, ion exchange particles and are designed for high resolution, high recovery and highly efficient separations of peptides, oligonucleotides and proteins.
- Uniform, densely packed ion exchange functional groups are chemically bonded to hydrophilic layer to increase column capacity
- Particles, coating and bonding are resistant to high pressures, promoting higher resolution and faster separations
- Multiple ion exchange groups are captured on one anchoring to increase capacity
It offers strong cation exchange (SCX), weak cation exchange (WCX), strong anion exchange (SAX) and weak anion exchange (WAX) phases. All phases are available in 1,7 µm , 3 µm, 5 µm and 10 µm non-porous particles sizes. Highly cross linked and rigid nonporous polystyrene divinylbenzene (PS/DVB) particles are grafted with a hydrophilic, polymeric layer, eliminating nonspecific binding.
- Catalog No:
- HEWL5190-2432
- HEWL5190-2472
- HEWL5190-2471
- HEWL5190-2467
- HEWL5190-2492
- HEWL5190-2464
- HEWL5190-2435
- HEWL5190-2463
- HEWL5190-2485
- HEWL5190-2480
- HEWL5190-2431
- HEWL5190-2479
- HEWL5190-2427
- HEWL5190-2473
- HEWL5190-2465
- HEWL5190-2491
- HEWL5190-2439
- HEWL5190-2487
- HEWL5190-2443
- HEWL5190-2440
- HEWL5190-2425
- HEWL5190-2453
- HEWL5190-2499
- HEWL5190-2424
- HEWL5190-2452
- HEWL5190-2495
- HEWL5190-2423
- HEWL5190-2451
- HEWL5190-2421
- HEWL5190-2444
- HEWL5190-2493
- HEWL5190-2461
- HEWL5190-2460
- HEWL5190-2459
- HEWL5190-2426
- HEWL5190-2454
- HEWL5190-2500
- HEWL5190-2494
- Max. pressure:
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 600 bar
- 400 bar
- 600 bar
- 600 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 600 bar
- 600 bar
- 400 bar
- 400 bar
- 400 bar
- 600 bar
- 400 bar
- 600 bar
- 600 bar
- 400 bar
- 600 bar
- 400 bar
- 400 bar
- 600 bar
- 400 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 400 bar
- 400 bar
- 600 bar
- 600 bar
- 400 bar
- 600 bar
- pH range:
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- Temperature limit:
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
Frequently Bought Together