Orders LinkContactUsLinkComponent
Micro pipettes, disposable, with ring mark
Micro pipettes, disposable, with ring mark
Micro pipettes, disposable, with ring mark
Certificates
Micro pipettes, disposable, with ring mark
Micro pipettes, disposable, with ring mark
4 Options Available Below
Product Details & Documents
Disposable micro pipettes with ring mark.
  • Calibrated to contain 'In'
  • Colour-coded
Certifications: DIN ISO 7550
Product Family Options
Product Information
  • Capacity
    Colour code
    Type
    Availability
    Price
  • 1/2/3/4/5 µl
    White
    Five marks
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 27,90 €
    27,90 €
    1 * 250 ST
       27,90 €
  • 25 µl
    2 × white
    One mark
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 26,20 €
    26,20 €
    1 * 250 ST
       26,20 €
  • 50 µl
    Green
    One mark
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 24,40 €
    24,40 €
    1 * 250 ST
       24,40 €
  • 100 µl
    Blue
    One mark
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 35,30 €
    35,30 €
    1 * 250 ST
       35,30 €