ContactUsLinkComponent
Counting chamber, Thoma new
Counting chamber, Thoma new
Catalog # HECH40447702
undefined
Counting chamber, Thoma new
Catalog # HECH40447702
Specifications
  • Depth of chamber:
    0,1 mm
  • Type:
    Thoma new
  • Description:
    Counting chamber with double ruling
  • Pk:
    1
Specifications
About this item
Thoma new counting chamber for blood tests and urine analysis.