Micro-V vials, CrossLab
Supplier: Agilent
Certificates
About this item
These vials are made from first hydrolytic glass type 1 and are available with screw, crimp or snap tops.
- Wide opening with low residual volume
- Compatible with 11 mm crimp closures
Frequently Bought Together