Order Entry
Austria
ContactUsLinkComponent
Dispersing elements (aggregates) for POLYTRON® PT 4000
Dispersing elements (aggregates) for POLYTRON® PT 4000
Catalog # 431-0205
Supplier:  KINEMATICA
undefined
Dispersing elements (aggregates) for POLYTRON® PT 4000
Catalog # 431-0205
Supplier:  KINEMATICA

Specifications

  • Speed:
    27 m/s
  • Volume:
    100 - 2500 ml
  • Description:
    PT-DA 20/2EC-E192
  • Pk:
    1
  • Stator/rotor Ø:
    15 / 20 mm
  • For unit:
    PT 4000

Specifications

About this item