ContactUsLinkComponent
Aerosol analysis monitors
Aerosol analysis monitors
Supplier:  Merck
Certificates
undefined
Aerosol analysis monitors
Supplier:  Merck
Specifications
  • Catalog No:
  • MILFMAWP025AC
  • MILFAP1002500
  • MILFMAWP025A0
  • Pore size:
  • 0,8/5,0 µm
  • 0,8/5,0 µm
  • Material:
  • PP
  • PS
  • Filtration area:
  • 4 cm²
  • 4 cm²
  • Ø:
  • 25 mm
  • 25 mm
  • Inlet/outlet connections:
  • Hose barb/-
  • Female Luer slip/female Luer slip