ContactUsLinkComponent
DIALYZER-Enhance™
DIALYZER-Enhance™
Catalog # 786-627
Supplier:  G-Biosciences
CAS Number:  
undefined
DIALYZER-Enhance™
Catalog # 786-627
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    50 mL
  • Description:
    DIALYZER-Enhance™, for improved dialysis time and rates
Specifications
About this item
For improved dialysis time and rates.
A product that when added to the dialysis buffer shifts the equilibrium resulting in the increased removal of a wide range of small molecules. The DIALYZER-Enhance™ consists of unreactive reagents that will not interfere or modify your reagents and will not cross the dialysis membrane, ensuring a pure, clean sample at the end of dialysis.