ContactUsLinkComponent
Beakers, low form
Beakers, low form
Catalog # RSGA070.005
Supplier:  RSG
Supplier #: 070.005
undefined
Beakers, low form
Catalog # RSGA070.005
Supplier:  RSG
Supplier #: 070.005
Log in for availability
Select an Option
180 mm
Select an Option
5000 ml
Select an Option
210 mm
Select an Option
07
Select an Option
1
About this item
Stainless steel 18/10, with spout.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.
Frequently Bought Together