ContactUsLinkComponent
Ion selective electrode system, AmmoLyt® Plus
Ion selective electrode system, AmmoLyt® Plus
Catalog # WTWO107042
Supplier:  WTW ONLINE
Ion selective electrode system, AmmoLyt® Plus
Catalog # WTWO107042
Supplier:  WTW ONLINE
Specifications
  • Type:
    VARiON® Ref
  • Description:
    Reference electrode, VARiON® Ref, for AmmoLyt® Plus/NitraLyt®Plus 700 IQ/VARiON® Plus
  • Pk:
    1
Specifications
About this item
This system – consisting of a robust, digital armature with ammonium replacement electrode and ammonium reference electrode – allows ammonium measurement directly in the medium, without sample preparation. The system is designed for connection to the IQ SENSOR NET multiparameter measuring system.
  • Calibration-free, long term stable
  • With integrated temperature sensor
  • Short response time
  • With screw/plug connector for connection of the SACIQ sensor connection cable
Ordering information: Supplied with unequipped sensor AmmoLyt® Plus 700 IQ, reference electrode VARiON® Ref, VARiON® Plus NH4 (ammonium electrode), special socket wrench, protective hood VARiON® 700 IQ-SK, Potassium chloride solution for storing the reference electrode and operating manual.