Beaker cups, medicine
Catalog # SARS75.1337.506
Supplier: SARSTEDT
Supplier #: 75.1337.506
Log in for availability
About this item
PP, translucent.
- Good temperature resistance
- Good chemical resistance
- Available in different colours
Ideal for general laboratory use and a wide variety of other applications. With embossed scale.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Frequently Bought Together