Orders LinkContactUsLinkComponent
Flow-through vessel
Supplier:   WTW
Flow-through vessel
Supplier:  WTW
Certificates
Flow-through vessel
Flow-through vessel
4 Options Available Below
Product Details & Documents
Flow vessels for WTW conductivity measuring cells are perfectly designed to perform measurements without the interference of carbon dioxide through the air or to examine the conductivity of solutions in closed circuits.
D 3Sen: Designed for everybody who must perform flow-through measurements in the field, i.e. ground water pumping tests. It consists of a rugged flow-through vessel in holder and a tripod, also suitable for pole mounting. With hose fittings for standard hoses with a diameter of 19 mm. Three places are available for pH/ORP, conductivity and dissolved oxygen sensors (CellOx/FDO). Including mounting accessories.
 : Please note: The overpressure in the system must not exceed 0,5 bar.
Product Family Options
Product Information
  • Availability
    Price
  • 715-2994
    Flow-through vessel, transparent PVC, i-Ø 18 mm, D 1/T, for old measuring cells
    Flow-through vessel, transparent PVC, i-Ø 18 mm, D 1/T, for old measuring cells
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 610,00 €
    610,00 €
    1 * 1 ST
       610,00 €
  • 663-1803
    Flow-through vessel, glass, D01/T, for LR 325/01
    Flow-through vessel, glass, D01/T, for LR 325/01
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 235,00 €
    235,00 €
    1 * 1 ST
       235,00 €
  • WTWA302760
    Replacement flow through vessel, D 001, for LR 325/001
    Replacement flow through vessel, D 001, for LR 325/001
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 369,00 €
    369,00 €
    1 * 1 ST
       369,00 €
  • WTWA903842
    Flow vessel for field measurements, D 3Sen
    Flow vessel for field measurements, D 3Sen
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 762,00 €
    762,00 €
    1 * 1 ST
       762,00 €