Reference electrodes, Radiometer Analytical
About this item
Radiometer Analytical reference electrodes are available with a variety of reference systems and liquid junctions.
- Liquid junction: porous pin
- Red-Rod technology provides a reproducible, fast and accurate response
- Calomel reference electrode can be used for special purposes
* These electrodes contain mercury, please dispose according to regulations.
** Screw cap electrodes require an external connection cable for connection to a meter.
4 Options Available Below
Product Details & Documents
Radiometer Analytical reference electrodes are available with a variety of reference systems and liquid junctions.
- Liquid junction: porous pin
- Red-Rod technology provides a reproducible, fast and accurate response
- Calomel reference electrode can be used for special purposes
* These electrodes contain mercury, please dispose according to regulations.
** Screw cap electrodes require an external connection cable for connection to a meter.
Product Family Options
Product Information
- Temp.TypeMaterialElectrolyteConnectionØ×LForAvailabilityPrice
- −5...+80 °CXR300, Ag/AgCl referenceGlassSat. KCl + sat. AgCl, refillableScrew cap**8×120 mmVoltammetry applicationsLog in for availability1 * 1 ST279,00 €
- −10...+100 °CREF251, red-rod referenceGlassSat. KCl, refillableBanana plug, fixed 1 m cable12×103 mmChallenging samples or general purpose measurementsLog in for availability1 * 1 ST779,00 €
- −5...+80 °CREF321, Ag/AgCl referenceGlass3M KCl + sat. AgCl, refillableScrew cap**8×103 mmGeneral or challenging samplesLog in for availability1 * 1 ST255,00 €
- −10...+100 °CREF201, red-rod referenceGlassSat. KCl, refillableBanana plug, 1 m fixed cable7,5×103 mmGeneral purposesLog in for availability1 * 1 ST361,00 €
Accessories
Product Information
- Catalog #LengthAvailabilityPrice
- Electrode connection cable, CL111, for connecting screw cap electrodes (S7) to banana instrument socketsElectrode connection cable, CL111, for connecting screw cap electrodes (S7) to banana instrument socketsHACHA94L1111 mLog in for availability1 * 1 ST - 69,20 €
Product Information
- Catalog #LengthAvailabilityPrice
- Electrode connection cable, CL114, for connecting screw cap electrodes (S7) to BNC instrument socketsElectrode connection cable, CL114, for connecting screw cap electrodes (S7) to BNC instrument socketsHACHA94L1141 mLog in for availability1 * 1 ST - 108,00 €