Orders LinkContactUsLinkComponent
ThermoScientific1920.png
Thermo Scientific Molecular Biology selection
Thermo Scientific Molecular Biology selection | Avantor
Top customer picks. Making a difference in labs worldwide
Thermo Scientific™ molecular biology tools are used by researchers worldwide. From quality high-fidelity or Taq DNA polymerases, simple and fast tools for cloning and electrophoresis, reliable cDNA synthesis products to specialised PCR enzymes, they are high quality essentials for everyday applications.
Here are a selection of kits that will ensure reliable performance in every step of the traditional molecular biology workflow including restriction enzymes, reverse transcription, PCR, and DNA ladders. As not all plastics are created equal, for maximum PCR performance also use Thermo Scientific PCR plastic consumables (tubes and tube strips, 96- and 384- well plates and sealing plate options).
Contact your VWR Life Science specialist to get more information about available samples.
ThermoScientificDNA.png
FastDigest Restriction Enzymes
176 restriction enzymes with 100% activity in one universal buffer. Also compatible with widely used downstream modifying enzymes.
Learn more
Maxima Reverse Transcriptases 540.png
Maxima Reverse Transcriptases
DirectPCRKits540.png
Direct PCR Kits
For DNA amplification from variety of unpurified samples
Ordering information
Download Brochure
GeneRulerDNAladders540.png
GeneRuler DNA ladders
For accurate DNA fragments analysis in agarose or polyacrylamide gels
Ordering information
Download Brochure
Selected Videos
Thermo Scientific Maxima™ First Strand cDNA Synthesis Kits with double-stranded DNase combine genomic DNA elimination and cDNA synthesis into a simple, one tube workflow that can be completed in as few as 15 minutes.