Orders LinkContactUsLinkComponent
bertin-hero.jpg
Improve Your Sample Preparation
Effective Tissue Disruption & Cell Lysis
In most Life Science workflows, one of the first steps is to disrupt your sample with a tissue homogeniser before the extraction of molecules of interest.
Discover numerous established Protocols
Improve Your Sample Preparation | Avantor
Using highly sophisticated bead-beating technology the Precellys® and Minilys® bench top homogenisers offer great benefits!
  • Highest yields: Efficient lysis and minimal sample loss - cooling option to protect sensitive target molecules
  • Time saving: Homogenisation takes seconds not minutes - process up to 24 samples in parallel
  • Reproducibility: Programmable and storable settings - no user fatigue or variance between individual samples
  • Contamination-free: Closed tube approach - no contact between sample and environment
  • Widest application range: Flexible grinding parameters - huge choice of optimised lysing kits in different tube sizes
Precellys Evolution – SUPER homogeniser
Precellys-Evolution-540.jpg
Precellys Evolution in Spotlight
The Precellys® Evolution offers optimal robustness, safety, efficiency and flexibility. Especially designed for Bio-safety class L3 laboratories, the Precellys® Evolution is based on 10 years of sample preparation expertise.
Free Precellys Lysing Kit Samples!
Precellys in action!
Product Selection
Precellys-Evolution-400.jpg
The most advanced tissue homogeniser offering widest application range and highest throughput.  
Learn more
Cooling-system-540.jpg
Cryolys® Evolution
Ensuring integrity of sensitive samples throughout homogenisation.
Learn more
Minilys-400.jpg
Minilys®
The cost- and space-saving solution for low throughput laboratory workflows.
Learn more
kits-400.jpg
Precellys® Kits
Tubes pre-filled with beads. Available in different volumes and bead sizes or blends to match any need.
Learn more
bertin_img.jpg
Extract & Purify
Also interested in solutions for nucleic acid purification? RNA, genomic DNA or plasmids from various sources? Clean-up of PCR or labelling reactions? Manual or automated sample processing?
Check out what VWR can offer you