Orders LinkContactUsLinkComponent
CamerasFeatureCard955x590.jpg
Cameras
Avantor offers the largest choice of cameras on the market. They cover most customers applications and market segments. If you are looking for an 'all in one' solution for microscopes, take a look at digital microscopes with an integrated camera.
Cameras | Avantor
CamerasColumnCardimg1.jpg
How to select!
As Avantor offers such a large range of cameras, you will find a selector to help you find the right one for you. The selector gives you the following choices:
Sensor type - Sensor size - Colour or monochrome - Resolution - Connectivity (USB or HDMI or other) - Application.
Camera selector
CamerasColumnCardimg2.jpg
USB cameras and cameras for fluorescence applications
The USB cameras are most often used. They are ideal for general purpose and high-end view. The USB cameras require a software for extended image analysis, which can be on a CD or to be downloaded from the website.
Models available from 3 to 20 MP
Go to the selector
CamerasColumnCardimg3.jpg
HDMI Cameras
The HDMI cameras offer several settings directly from the device by using a mouse (generally supplied) to select the preferred parameters.
Some models can be used with a software (via USB connection to PC) With HDMI cameras you can show pictures & videos on a beamer or external monitor.
From 2MP to 6MP
Go to the selector
CamerasColumnCardimg4.jpg
WiFi and tablet cameras
Tablet cameras have an application on an iPad, or can use application software, depending on the type of camera. It is easy to share images. No PC is needed. Ideal for basic image analysis.
5 MP models.
Go to the selector
CamerasColumnCardimg5.jpg
Eyepiece cameras
For basic applications, they can be easily added to a microscope, when there is not a phototube or a trinocular microscope.
Eyepiece camera
Advanced DinoLite eyepiece camera