Orders LinkContactUsLinkComponent
reinsmediahero1960x760.png
Process chromatography resins and media
Avantor J.T.Baker® brand BAKERBOND® process chromatography products feature a unique multi-mode chemistry designed to increase yield by improving the efficiency of your purification process and enhance selectivity by removing closely related impurities. Our resins and media are proven to reduce total purification steps while also improving purity and yield in bioprocessing applications.
asolution.png
A solution for each step in the process
The J.T.Baker® brand BAKERBOND® process chromatography offering includes a range of trusted products, suitable for the protein A capture step in mAbs manufacturing, through polishing steps in mAbs, vaccine production, recombinant protein production and more.
Browse products
j-t-baker-logo-200x200.png
A legacy of excellence in the most demanding applications
For more than a century, customers around the world have trusted Avantor J.T.Baker® brand products to meet the needs of some of the most demanding applications.
Coupled with our trusted offering of cGMP chemicals and excipients, J.T.Baker® brand process chromatography media, columns and resins offer exacting performance in the critical chromatography steps in mAbs downstream processing.
Browse our range of cGMP chemicals and excipients from J.T.Baker® and other brands.
Browse our range of cGMP chemicals and excipients
Don’t miss these insights
scale-up-development-services-540x390.jpg
With over four decades of experience in chromatography, our team of experts can assist with a range of support—from column packing to custom-made media. We also offer our expertise in downstream process optimization, working with you to recommend the proper selection of buffer cleaning and solutions for your application.
Learn more about our downstream process optimization services.
mabs-downstream-purification-540x390.jpg
Increased Viral Clearance in Downstream Process Manufacturing Through Use of Novel Purification Resins and Additives.
Watch Now
Product selection
J.T.Baker® BAKERBOND® PROchievA™ Resin
Recombinant protein A resin offering superior dynamic binding capacity with excellent alkaline stability in mAbs manufacturing.
J.T.Baker® BAKERBOND® PolyPEI bulk resin
Multimode ion exchange (IEX) chromatography resin to enhance the separation power of your chromatography process steps.
Multimode ion exchange (IEX) chromatography resin with unique selectivity to enable higher purification performance.
J.T.Baker® BAKERBOND® Poly Hi-Propyl bulk resin
Mixed mode hydrophobic interaction chromatography (HIC) resin.
J.T.Baker® BAKERBOND® PolyQUAT bulk resin
Multimode ion exchange (IEX) chromatography resin to enhance the separation power of your chromatography process steps.
Multimode ion exchange (IEX) chromatography resin for process chromatography.
Suitable for use in mAbs manufacturing with a range of common chromatography instruments in convenient 1ml and 5ml lab columns.
J.T.Baker® BAKERBOND® PolyPEI™ resin in pre-packaged lab columns
Multimode ion exchange (IEX) chromatography resin in convenient 1ml and 5ml lab columns.
J.T.Baker® BAKERBOND® PolyCSx™ resin in pre-packaged lab columns
Multimode ion exchange (IEX) chromatography resin in convenient 1ml and 5ml lab columns.
J.T.Baker® BAKERBOND® Poly Hi-Propyl™ resin in pre-packaged lab columns
Mixed mode hydrophobic interaction chromatography (HIC) resin in convenient 1ml and 5ml lab columns.
J.T.Baker® BAKERBOND® PolyQUAT™ resin in pre-packaged lab columns
Mixed mode hydrophobic interaction chromatography (HIC) resin in convenient 1ml and 5ml lab columns.
Related content
Mixed mode hydrophobic interaction chromatography (HIC) resin.
Multimode strong cation exchange chromatography resin.
J.T. Baker® BAKERBOND® PolyPEI multimode weak anion exchange chromatography resin.
J.T. Baker® BAKERBOND® PolyQUAT multimode strong anion exchange chromatography resin.
J.T.Baker® BAKERBOND® PolyABx Multimode Weak Cation Exchange Chromatography Resin
J.T.Baker® Binding affinity of PROchievA resin to IgG from different species
J.T.Baker® Specific binding to Fc domain of BAKERBOND® PROchievA resin
J.T.Baker® BAKERBOND® PROchievA column packing guidelines
Avantor J.T.Baker® brand BAKERBOND® process chromatography resins comparison guide
Ready to move science forward?
We’re here to help.
Yes, let's get started!