Orders LinkContactUsLinkComponent
qpcr-hero1920x760.jpg
qPCR
qPCR stands for Quantitative Real-Time Polymerase Chain Reaction and is a technique which monitors, in real-time, the enzymatic amplification of a targeted sequence of DNA. Analysis of the resultant amplification curves informs a researcher of the starting amount of their DNA. Avantor® offers a full range qPCR solutions to meet your needs.
qPCR | Avantor
Featured Suppliers
beckman.jpg
Supporting nucleic acid extraction, PCR/qPCR/ddPCR and enzymatic post-reaction clean-ups.
Agilent-Molecular-Biology-Tools.jpg
Agilent’s tools span from innovative genome engineering to precision tools and solutions that are customizable.
Extraction
extraction_540x390.jpg
Extraction reagents and homogenizers for sample preparation
See our phenol:cloroform extraction reagents.
Choose from our selection of Proteinase K reagents.
Manual or automated benchtop homogenizers have user-friendly control systems for straightforward processing.
Purification
purification.jpg
Purification process to get nucleic acid samples ready for downstream application.
Get the automated nucleic acid purification systems you need for your lab.
Reagents and kits for DNA purification.
Buffers and systems for nucleic acid stability during sample storage/transport.
RNA purification solutions including reagents, kits and accessories.
Reagents and kits to isolate plasmid DNA.
Filter plates are essential for biomolecule purifications and separations.
See our selection of optimized devices for the vacuum filtration of filter plates.
Designed for small scale separations between liquid and magnetic beads.
Reverse Transcription
qpcr_reverse_transcription.jpg
cDNA synthesis from RNA.
Reagents and kits to synthetize cDNA.
Reagents and kits for RT-qPCR.
See our selection of one-step probe reagents for RT-qPCR.
Selection of cleaning agents to remove, inactivate, or completely destroy contaminants.
PCR workstation enclosures provide the contamination-free environment required for the most sensitive DNA segment detection.
Amplification
qpcr_amplification.jpg
DNA sample is added to reaction mix including DNA polymerase, nucleotides, primers and amplification reactions are done in a thermal cycler.
Master Mixes provide everything needed to perform qPCR. Just add template and primers and start amplifying.
See our thermal cyclers for qPCR applications.
View our wide selection of high-purity Nucleotides.
We offer an extensive variety of PCR Plates.
Broad range of microplate sealing films and foils to reduce evaporation during reactions.
Discover our large selection of high-quality PCR tubes.
Filters will block aerosol contaminants to protect both the equipment and sample.
undefined
Questions about our offering of qPCR products?
Contact us today to discuss your needs. Our team is ready to help.
Contact us
Are you ready to move your next innovation forward? We can help.