Orders LinkContactUsLinkComponent
Endonuclease | Avantor
Endonuclease
Featured Solutions
endo-Recovered.jpg
DENARASE® is a highly active endonuclease from Serratia marcescens, it is widely used in biopharmaceutical manufacturing to quantitatively eliminate host cell DNA and RNA during the production of biologicals and vaccines.