Orders LinkContactUsLinkComponent
Centrifuges | Avantor
Centrifuges
Featured Solutions
Micro-Star1.jpg
Micro Star 17 and 17R microcentrifuges combine power, versatility and convenience in a safe, compact, easy to use laboratory instrument. Both the ventilated Micro Star 17 and the refrigerated Micro Star 17R are designed to accelerate the routine sample preparation process for ie. PCR!
Additional Resources
Microcentrifuges, ventilated/refrigerated, Micro Star 17 / 17R
BENCHTOPMICRO.jpg
These small bench top centrifuges are ideal for sample prep in biotechnical industries, as well as for academic research. Flexibility to spin ie. 50mL conical tubes at over 12,000 x g or a wide range of microtubes at speeds up to 30,279 x g
Additional Resources
Centrifuges, bench top, ventilated/refrigerated, Mega Star 600 / 600R
versatile-centrifuges.jpg
These versatile centrifuges are ideal for a range of routine applications in genomics/molecular biology, proteomics and biochemistry. Create your perfect solution in seconds.
Additional Resources
Centrifuges, bench top, ventilated/refrigerated, Mega Star 1.6 / 1.6R