ContactUsLinkComponent
bimos_bildnr-6636_vwrBlueline_2017-12-12-955x900.jpg
VWR® ergonomic chairs for laboratory and industrial environments
Sitting for prolonged periods in front of your microscope, pipetting at your bench or just using your computer can cause fatigue, circulatory disorders and posture problems. VWR® ergonomic chairs and stools help keep you and your staff healthier and more productive.
The human body is not optimized for long stretches of sitting. But when we need to, using ergonomically designed chairs and stools can keep us healthy and productive. Choose from the VWR® smart ergonomic chair and stool portfolio for your operations.
Economy_Line2_teaser_140_140.jpg
Our Economy Line 2.0 is a simple laboratory chair that meets all laboratory work requirements. It features reliable body support thanks to a permanent contact backrest. Seat height and backrest adjustment deliver unbeatable price and performance.
Find helpful resources
Explore more
Mr_La2_seitlich_teaser_140_140.jpg
Mr. Lab 2.0 features proven laboratory ergonomics, hygienic upholstery and innovative airflow technology for ideal climate comfort. Auto-synchron technology with automatic weight regulation and smart details, such as the integrated hook and labelling field, offers superior performance and value.
Find helpful resources
Explore more
professional-chair-140.jpg
Engineered after two years of study with ergonomic experts and designers, these professional-level chairs are auto adaptive, so no adjustment is needed. In addition, body weight is managed evenly, so there is a reduction of pressure under the leg.
Find helpful resources
Explore more
132-0909_140.jpg
Neon chairs are designed for busy users and feature sustainability and economy. If the upholstery wears out, it can be easily replaced. These chairs combine ergonomics and functional characteristics with a variety of upholstery options, including the super-comfortable SUPERTEC fabric.
Find helpful resources
Explore more
132-0421_140.jpg
Cleanrooms and sterile environments across multiple industries present unique seating challenges. Our cleanroom chairs combine comfort while meeting the requirements of key air cleanliness classification 3 by DIN EN ISO 14644-1 and EU GMP guideline specifications.
Find helpful resources
Explore more
132-0419_140.jpg
ESD chairs are for use in sensitive areas requiring discharge resistance per EN 61340-5-1. These low-cost chairs feature an ergonomic design and a reliable discharge protection system. 
Find helpful resources
Explore more
132-0425_243.jpg
Laboratory stools with a black artificial leather seat and adjustable height with a pneumatic spring.
VWRI132-0963_243.jpg
Laboratory stools with an extra-wide  seat in black skai® PU foam and PA base (plastic and black steel).
132-1297_243.jpg
Laboratory stools with a large seating surface and upholstery made from black PU foam.
VWRI132-1099_243.jpg
VWR ergonomic stools feature black PU foam seats and automatically adapt to users, facilitating better posture and minimizing spinal stress.
132-0424_243.jpg
Stools that meet the requirements for cleanliness classification 3 in accordance with DIN EN ISO 14644-1 as well as other standards.
Watch more vidoes
23481408_th.jpg
See how VWR® Blue Line Seating automatically ensures good seating comfort.
23481413_th.jpg
See how a simple chair feature can help users find their chair with ease.
Request a quote
Our team of experts is ready to help you select the best VWR® ergonomic seating for your needs.
Request a quote